Learn More
Ace Glass, Inc. O-RING CAPFE SIZE 228 O-RING CAPFE SIZE 228
SDP
Supplier:  Ace Glass, Inc. 7855847
Encompass_Preferred
Size 228 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-975-726