Learn More
Ace Glass, Inc. O-RING EPDM SIZE 114 O-RING EPDM SIZE 114
SDP
Supplier:  Ace Glass, Inc. 7855922
Encompass_Preferred
Size 114 Ethylene Propylene (EPDM) o-ring. Pack/12. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.
Catalog No. 50-979-569