Learn More
Ace Glass, Inc. O-RING FETFE SIZE 226 O-RING FETFE SIZE 226
SDP
Supplier:  Ace Glass, Inc. 7855746
Encompass_Preferred
Size 226 FETFE o-ring, Pack/3. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-979-553