Learn More
Glass Expansion O-ring Kit for Eluo (2 sets)
SDP
Supplier:  Glass Expansion 700806
Encompass_Preferred
O-Ring Kit for Eluo (2 Sets). An O-Ring kit providing two changes of O-rings for the popular Eluo Nebulizer Cleaning Tool (NC9462559, 70-ELUO). Use O-ring removal tool 50-100-1636.Try using Fluka RBS-25 Concentrate 1L (NC9000117, FLUKA25) with your Eluo, it is a concentrated cleaning solution that is highly effective in removing clogs from nebulizers, and deposits/residues from spray chambers, torches, injectors, and cones.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1321457