Learn More
Beckman Coulter Beckman Coulter O-Ring for HIAC sensor fittings (808-010-200)
Supplier:  Beckman Coulter 808010200
Encompass
Replacement O-ring for sensor fittings used on Beckman Coulter liquid particle counters such as the HIAC 9703+. The O-ring provides a sealing interface for sensor plumbing and helps prevent leaks and contamination in particle-counting sample lines.
  • Replacement o-ring for sensor fittings.
  • Provides a leak-resistant seal for sensor connections.
  • Viton elastomer for chemical and temperature resistance.
  • Sold as a pack; manufacturer's replacement parts list shows pack of 4.
  • Helps maintain sensor performance and reduce service interruptions.
Catalog No. NC3312043