Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 230 O-RING KALREZ SIZE 230
SDP
Supplier:  Ace Glass, Inc. 7855689
Encompass
Size 230 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-979-546