Learn More
Chemglass Life Sciences O-Ring, FEP-Silicone, 100mm Flange
SDP
Supplier:  Chemglass Life Sciences CG14741
Encompass_Preferred
O-Ring, FEP-Silicone, 100mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200°CSilicone: -50 to 230°CPerfluoro: -7 to 230°CFEP Encapsulated Silicone: -62 to 205°CFEP Encapsulated Stainless: -250 to 205°C
Catalog No. CG14741