Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-ring, Size #113, White
SDP
Supplier:  Chemglass Life Sciences CG309U113
Encompass_Preferred
Chemglass Perfluoro (FFKM) O-ring #113 is a thermally stable, chemically resistant sealing element designed for demanding laboratory and process applications. Manufactured from perfluoroelastomer, it provides superior resistance across a wide range of aggressive chemicals and elevated temperatures.
  • Perfluoroelastomer (FFKM) construction for exceptional chemical resistance.
  • Wide service temperature range: -7 to 230° C.
  • White 75 durometer stock for durable sealing performance.
  • Compatible with acids, inorganic bases, alcohols, ethers, hydrocarbons, esters, aldehydes, ketones, and amines.
  • Sized for common laboratory fittings and fluid-handling components.
Catalog No. 50-274-3567