Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #217, White
SDP
Supplier:  Chemglass Life Sciences CG309U217
Encompass_Preferred
The Chemglass perfluoro (FFKM) O-ring, size #217 in white, is a high-performance perfluoroelastomer seal engineered for demanding chemical and thermal environments. It delivers superior chemical resistance and thermal stability for reliable sealing in laboratory fittings, valves, and reactors.
  • Provides effective service temperature range of -7 to 230° C
  • Resists a wide range of aggressive chemicals
  • 75 durometer hardness for reliable sealing
  • White color for easy identification during maintenance
  • Suitable for valves, fittings, reactors, and general laboratory equipment
Catalog No. 50-274-3549