Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), Size 223, White
SDP
Supplier:  Chemglass Life Sciences CG309U223
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring, size 223, white, offers thermally stable, high-chemical-resistance sealing for laboratory and process equipment. It is intended for use in corrosive or high-temperature environments where broad solvent compatibility is required.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color with 75 durometer typical hardness.
  • Size 223 sealing profile for standard O-ring fittings and joints.
  • Effective service temperature range of -7 to 230° C.
  • Compatible with acids, bases, alcohols, ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Suitable for valves, fittings, vacuum components, and glassware seals.
  • Cost-effective perfluoroelastomer option for demanding chemical applications.
Catalog No. 50-274-3544