Learn More
Ace Glass, Inc. O-RING VITON SIZE 108 O-RING VITON SIZE 108
SDP
Supplier:  Ace Glass, Inc. 785511
Encompass_Preferred
Size 108 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-979-496