Learn More
Medchemexpress LLC o-Toluic acid-13C | 70838-82-3 | 977702 | 99% | 137.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY41494S150MG
Encompass_Preferred
o-Toluic acid-13C is the 13C labeled o-Toluic acid, which is a benzoic acid substituted by a methyl group at position 2. It plays a role as a xenobiotic metabolite.
  • Isotope-labeled compounds
  • Endogenous metabolite
  • Stable heavy isotope of carbon (13C)
  • Appearance: solid
  • Color: white to off-white
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-04579