Learn More
Medchemexpress LLC OAB-14, a bexarotene derivative | 2140911-49-3 | 99.2% | C32H46N4O2 | 10MG

Supplier: Medchemexpress LLC HY13997310MG
OAB-14 is a bexarotene-derived research compound investigated for promoting amyloid-β clearance and improving Alzheimer's disease-related pathology. It is provided as a purified powder with documented storage recommendations for powder and solutions to preserve stability.
- Used in preclinical studies targeting amyloid-β clearance.
- High purity suitable for research applications.
- Molecular formula C32H46N4O2 and molecular weight 518.73.
- Available in small milligram pack sizes for laboratory experiments.
- Storage conditions provided for powder and in solvent to maintain stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.