Learn More
Cole-Parmer Oakton pH/mV Simulator, USA-NIST-Custom mV Output
Supplier:  Cole-Parmer 3565200
Encompass_Preferred
Check for meter errors and ensure properly calibrated equipment with this pocket-sized pH simulator. Simple push-button operation allows for selection of USA buffers, NIST buffers, and mV output on the LED display. Additional features include 1 GΩ high impedance test, BNC connector, and low-battery indicator. Order protective rubber boot which serves as a benchtop stand separately.
Catalog No. NC9012084