Learn More
Selleck Chemical LLC O-Acetyl-L-serine hydrochloride S3348-25mg
SDP
Supplier:  Selleck Chemical LLC S334825MG
Encompass_Preferred
O-Acetyl-L-serine (OAS O-Acetylserine O-Acetyl-L-serine) hydrochloride (HCl) is an intermediate in the biosynthesis of the amino acid cysteine in bacteria and plants that displays a signalling function leading to changes in transcript levels of a specific gene set irrespective of the sulfur status of the plant
Catalog No. 50-310-8572