Learn More
Waters Corp Oasis HLB Online Column, 80Å, 15 µm, 4.6 mm X 20 mm, 1/pk
Supplier:  Waters Corp 186002044
Encompass_Preferred
The Oasis HLB on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE. The on-line column contains the universal polymeric Oasis HLB sorbent, which is optimized for the extraction of a wide range of analytes from various matrices. The Oasis HLB on-line column is designed for use with the appropriate Waters narrow-bore analytical column, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry column. The column features traditional HPLC column fittings and hardware.
Catalog No. 50-780-900