Learn More
Waters Corp Oasis Online Column, HLB, 80Å, 25 μm, 2.1 mm x 20 mm
Supplier:  Waters Corp 186002036
Encompass_Preferred
This online column is engineered for rugged, reproducible, and ultra-fast solid phase extraction (SPE). It features a universal polymeric sorbent optimized for extracting a broad range of analytes from various matrices, intended for use with appropriate narrow-bore analytical columns.
  • Rugged, reproducible, and ultra-fast on-line analysis with traditional HPLC column fittings.
  • Water-wettable sorbent eliminates equilibration and conditioning steps, reducing solvent consumption by up to 70% and sample preparation time by 40%.
  • Provides high recovery and reproducible results for a wide range of compounds.
Catalog No. 50-780-892