Learn More
Waters Corp Waters Oasis MCX Vac Cartridge, 6 cc, 150 mg Sorbent, 30 μm
Supplier:  Waters Corp 186000256
Encompass_Preferred
The 6 cc cartridges contain a novel, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups. This sorbent is water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need to prevent sorbent beds from drying out before sample loading.
  • Novel, mixed-mode polymeric sorbent
  • Optimized for higher selectivity and sensitivity for extracting basic compounds
  • Contains cation-exchange groups
  • Water wettable, maintaining retention and recoveries even if sorbent runs dry
  • Eliminates the need to prevent sorbent beds from drying out during critical steps prior to sample loading
Catalog No. 50-818-609