Learn More
Waters Corp Waters Oasis Sorbent Selection Flangeless Cartridge Kit, 30 mg/1 mL, Mixed-Mode Chemistries, 40 pk
Supplier:  Waters Corp 186006345
Encompass_Preferred
The Oasis sorbent selection flangeless cartridge kit is specifically designed for the rapid development of Solid Phase Extraction (SPE) methods for Liquid Chromatography-Mass Spectrometry (LC/MS) analysis. It contains 40 cartridges, with 10 each of four different Oasis mixed-mode, ion-exchange chemistries: MAX (strong anion exchanger), WAX (weak anion exchanger), MCX (strong cation exchanger), and WCX (weak cation exchanger). This kit facilitates quick and easy sample preparation method development using only two SPE protocols with these chemistries.
  • Rapid method development for SPE methods in LC/MS analysis.
  • Comprehensive sorbent selection with 40 cartridges, including four mixed-mode, ion-exchange chemistries: MAX, WAX, MCX, and WCX.
  • Simplified protocol for efficient sample preparation method development using only two SPE protocols.
Catalog No. 50-201-610