Learn More
Medchemexpress LLC Obacunone | 751-03-1 | 454.51 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN042810MM/1ML
Encompass_Preferred
Obacunone is a highly oxidized triterpenoid limonoid isolated from citrus plants. It exerts its anticancer effects by inducing apoptosis and protects retinal pigment epithelial (RPE) cells from ultraviolet (UV) radiation (UVR)-induced oxidative damage. This product is an analytical standard intended for research and analytical applications.
  • Inhibits UVR-induced cytotoxicity
  • Improves UVR-induced oxidative damage in ARPE-19 cells
  • Slows UVR-induced apoptosis in human ARPE-19 cells and murine RPE cells
  • Inhibits bleomycin-induced lung inflammation and fibrosis
  • Activates the Nrf2-dependent antioxidant responses
Catalog No. 50-003-88931