Learn More
Medchemexpress LLC Obacunone | 751-03-1 | 454.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0428R100MG
Encompass_Preferred
Obacunone (Standard) is the analytical standard for Obacunone, designed for research and analytical applications. It is a highly oxidized triterpenoid limonoid, naturally isolated from various citrus plants. Obacunone demonstrates anticancer properties primarily by inducing apoptosis and offers protective effects against ultraviolet (UV) radiation-induced oxidative damage in retinal pigment epithelial (RPE) cells.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Commonly employed in techniques such as HPLC, GC, and MS.
Catalog No. 50-003-88932