Learn More
Medchemexpress LLC OBCAM/OPCML Rat HE 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77115100UG
Encompass_Preferred
OBCAM/OPCML Protein binds opioids in acidic lipid environments influencing cellular communication and facilitating cell contact potentially impacting various biological processes OBCAM/OPCML Protein Rat (HEK293 Fc) is the recombinant rat-derived OBCAM/OPCML protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-86078