Learn More
Medchemexpress LLC Obeticholic acid | 459789-99-2 | ≥98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12222R100MG
Encompass_Preferred
Obeticholic acid is a potent, selective, orally active farnesoid X receptor (FXR) agonist used as a research reagent. It has anticholeretic and anti-inflammatory activity and is used to study FXR signaling and metabolic pathways.
  • Potent, selective FXR agonist with reported EC50 ≈ 99 nM.
  • High purity, ≥98.0% (1H NMR), white to off-white solid.
  • Identified by CAS 459789-99-2 and formula C26H44O4 (MW 420.63).
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-31364