Learn More
TARGETMOL CHEMICALS INC Obeticholic Acid 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T178950MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Obeticholic Acid (6-ECDCA, INT-747) is a high-affinity, semisynthetic, bile acid-derived FXR agonist with an EC50 of 99 nM and is able to upregulate IkB-alpha, KLF-2, and KLF-4 expression. Obeticholic Acid (6-ECDCA, INT-747) also shows potential for treating hepatic steatosis, inflammation, and fibrosis while increasing insulin sensitivity. Purity 99.78%
Catalog No. 50-287-8401