Learn More
Medchemexpress LLC Obeticholic acid-d4-1 | 459789-99-2 | 99.8% | 410.62 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12222S11MG
Encompass_Preferred
Obeticholic Acid-d4 is the deuterium labeled Obeticholic acid. Obeticholic acid (INT-747) is a potent, selective and orally active FXR agonist with an EC50 of 99 nM. Obeticholic acid has anticholeretic and anti-inflammation effect. Obeticholic acid also induces autophagy.
  • It is a deuterium labeled compound.
  • It is a potent, selective, and orally active FXR agonist.
  • It has an EC50 of 99 nM.
  • It possesses anticholeretic effects.
  • It exhibits anti-inflammation effects.
  • It induces autophagy.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37300