Learn More
Medchemexpress LLC Obeticholic acid-d5 | 1992000-80-2 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY12222S500UG
Encompass_Preferred
Deuterium-labeled obeticholic acid used as an internal standard for analytical and pharmacokinetic studies. Stable-isotope analog with formula C26H39D5O4 and molecular weight 425.66 g/mol; supplied as a 500 UG quantity.
  • Deuterium-labeled internal standard for LC-MS quantification.
  • Provides accurate mass discrimination for pharmacokinetic studies.
  • Supplied as a low-mass, ready-to-use reference material.
  • Suitable for method development and validation in bioanalysis.
Catalog No. 50-002-32656