Learn More
Medchemexpress LLC OBI-992 drug-linker | 3039487-10-7 | 96.6% | 2173.46 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1684895MG
Encompass_Preferred
OBI-992 drug-linker is a drug-linker conjugate composed of a TOP I inhibitor Exatecan and a linker. It exhibits antitumor property in mouse models and can be used for the synthesis of ADC OBI-992.
  • Appears as a solid.
  • Color is white to yellow.
  • Exhibits antitumor property in mouse models.
  • Can be used for the synthesis of ADC OBI-992.
Catalog No. 50-003-90398