Learn More
Medchemexpress LLC Obicetrapib | 866399-87-3 | 99.0% | 722.60 | C32H31F9N4O5 | 10 MG

Supplier: Medchemexpress LLC HY1877810MG
Obicetrapib is a selective cholesteryl ester transfer protein (CETP) inhibitor supplied as a research reagent for preclinical and biochemical studies. The compound is provided as a solid powder with high purity and is characterized by CAS 866399-87-3, molecular formula C32H31F9N4O5, and molecular weight 722.60 g/mol. Store under recommended conditions to maintain stability.
- Selective CETP inhibitor for lipid metabolism research.
- High purity compound (approximately 99.0%).
- White to light yellow solid powder suitable for small-scale assays.
- Molecular weight 722.60 g/mol and formula C32H31F9N4O5.
- CAS number 866399-87-3 for unambiguous identification.
- Packaged as 10 mg for preparative or analytical use.
- Storage: powder -20°C for long-term, 4°C for short-term; in solvent -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.