Learn More
Medchemexpress LLC Ocadusertib | 2382811-41-6 | 99.6% | 459.50 g·mol⁻¹ | C25H25N5O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14829710MG
Encompass_Preferred
Ocadusertib is a potent, selective inhibitor of receptor-interacting serine/threonine-protein kinase 1 (RIPK1) supplied as a high-purity small-molecule research compound for biochemical and cell-based studies. It blocks RIPK1-mediated necroptosis and inflammatory cell death and has demonstrated anti-necroptosis activity in TNFα-induced mouse L929 cells (EC50 < 1 nM).
  • Selective RIPK1 inhibitor mechanism of action.
  • Demonstrated anti-necroptosis activity (EC50 < 1 nM) in TNFα-induced L929 cells.
  • High purity: 99.57%.
  • Molecular weight 459.50 g·mol⁻¹ and formula C25H25N5O4.
  • Provided in small quantities for preclinical in vitro and cell-based assays.
Catalog No. 50-002-87139