Learn More
Enzo Life Sciences Ochratoxin A (25mg). CAS: 303-47-9
SDP
Supplier:  Enzo Life Sciences ALX630089M025
Encompass_Preferred
Mycotoxin. Natural contaminant of mouldy food and feed. It has a number of toxic effects, the most prominent being nephrotoxicity. Immunosuppressive, genotoxic, teratogenic and carcinogenic. Stimulates lipid peroxidation. Induces oxidative DNA lesions coupled with direct DNA adducts via quinone formation. Purity: ≥98% (HPLC, TLC). Appearance: White to off-white crystalline powder. Solubility: Soluble in methanol (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9618