Learn More
Medchemexpress LLC Ochratoxin B | 4825-86-9 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN67861MG
Encompass_Preferred
Ochratoxin B is an orally active secondary metabolite produced by *Aspergillus ochraceus*. It is a non-chlorinated analog of the mycotoxin Ochratoxin A. This compound has been shown to reduce the toxic effects of Ochratoxin A, inhibit cell division, and cause craniofacial malformations in *Xenopus laevis* embryos.
  • Orally active secondary metabolite.
  • Reduces toxic effects of Ochratoxin A.
  • Inhibits cell division.
  • Causes craniofacial malformations in Xenopus laevis embryos (though with less potency than Ochratoxin A).
  • Available in high purity (99.90%).
  • More extensively metabolized and rapidly eliminated than Ochratoxin A in rats.
Catalog No. 50-004-17968