Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ocifisertib fumarate | 1616420-30-4 | >98.0% | 650.7 g/mol | C37H38N4O7 | 50MG
SDP

Supplier:  Medchemexpress LLC HY12300B50MG

Encompass_Preferred

Ocifisertib fumarate is the fumarate salt of CFI-400945, a potent, selective, and orally bioavailable inhibitor of polo-like kinase 4 (PLK4) supplied for research use in biochemical and cellular studies of PLK4-related pathways and antineoplastic activity.

  • Fumarate salt form of the PLK4 inhibitor.
  • Potent biochemical activity: PLK4 Ki = 0.26 nM; PLK4 IC50 = 2.8 nM.
  • Molecular formula C37H38N4O7 and molecular weight 650.7 g/mol.
  • High purity (>98.0% by HPLC) suitable for research applications.
  • Supplied in small research pack sizes, including 50 mg.

Catalog No. 50-002-35727


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.