Learn More
Selleck Chemical LLC Oclacitinib E4971-5MG
SDP
Supplier:  Selleck Chemical LLC E49715MG
Encompass_Preferred
Oclacitinib(PF-03394197) an inhibitor of JAK is highly potent at inhibiting JAK1 with an IC50 value of 10 nM respectively Oclacitinib is highly potent against the JAK1 enzyme displaying a 1 8-fold selectivity for JAK1 vs JAK2 and a 9 9-fold selectivity toward JAK1 vs JAK3
Catalog No. 50-000-42710