Learn More
Lonza Walkersville OCP– Osteoclast Precursor Basal Medium
Supplier:  Lonza Walkersville PT8201
Encompass
OCP Octeoclast Precursor BulletKit Medium (provides both negative control/experimental medium which can be used as a negative control [without the addition of additional supplements] or an experimental medium [with the addition of researcher provided differentiation supplements] and a positive control medium guaranteed to induce differentiation into mature, functionally active osteoclasts; Each bottle of the OCPBulletKit Medium contains enough differentiation medium to fully differentiate approximately 1.5 million osteoclast precursor cells [approximately 1.5 complete 96-well plates] into mature, functionally active osteoclasts).
Catalog No. NC1950044