Learn More
Medchemexpress LLC Octacosane-d58 | 16416-33-4 | MFCD00190493 | 99.1% | 453.12 | C28D58 | 5 MG

Supplier: Medchemexpress LLC HYW272217S5MG
Octacosane-d58 is a deuterium-labeled octacosane used as an analytical standard and research reagent for studies of hydrocarbon metabolites. It is supplied as a high-purity, stable isotope-labeled powder suitable for tracer and analytical applications.
- Deuterium-labeled hydrocarbon for analytical and tracer use.
- Molecular formula C28D58.
- Molecular weight 453.12 g·mol-1.
- Purity 99.05%.
- Packaging 5 mg powder.
- Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
- Synonyms: n-Octacosane-d58, NSC 5549-d58.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.