Learn More
Cayman Chemical OctacosnoIc ACD mthyl Ste 10mg
SDP
Supplier:  Cayman Chemical 2672610 MG
Encompass_Preferred
A very long-chain fatty acid methyl ester; has been found in transesterified palm oil, biodiesel produced by the microalga Botryococcus, and in sediment samples from Lake Kivu in the EAst African rift valley
Catalog No. 50-293-6337