Learn More
Abcam 5-(Octadecanoylamino)fluorescein, 100MG
Supplier:  Abcam AB275323100MG
Encompass_Preferred
MW 613.8 Da, Purity >98%. Amphiphilic fluorescent probe useful in monitoring membrane lipid behaviour and fluorescence resonance energy transfer applications in life cells. The label makes the cell membrane bright green, Em/Ex: 520/490 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-204