Learn More
Medchemexpress LLC Octadecanoic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl ester | 79408-85-8 | MFCD00057311 | 99.0% | 442.63 g·mol⁻¹ | C28H42O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW017657100MG
Encompass_Preferred
4-Methylumbelliferyl stearate is a fluorogenic biochemical reagent used as an esterase and lipase substrate; enzymatic hydrolysis liberates fluorescent 4-methylumbelliferone for sensitive activity assays. It is supplied as a white to off-white solid with documented purity and stability for laboratory use.
  • Fluorogenic substrate for esterase and lipase assays.
  • Releases 4-methylumbelliferone upon enzymatic hydrolysis.
  • High purity for reproducible assay performance.
  • Available in multiple laboratory pack sizes to suit assay needs.
  • Stable at room temperature for long-term storage; in solution store frozen for extended stability.
Catalog No. 50-003-01818