Learn More
Biotium Octadecyl Rhodamine B
Supplier:  Biotium 60033
Encompass_Preferred
The fluorescence of octadecyl rhodamine B in membranes is quenched at high dye concentration but is released at dilution. This property makes the dye useful for membrane fusion assays. Properties: Ex/Em (MeOH) = 556/578 nm; Dark solid soluble in EtOH or DMSO; Store at -20°C and protect from light; C46H67ClN2O3; MW: 731.50; [65603-19-2].
Catalog No. 50-196-4727