Learn More
Medchemexpress LLC Octadecyl rhodamine B chloride | 65603-19-2 | MFCD00467892 | 98.7% | 731.49 g/mol | C46H67ClN2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW12770310MG
Encompass_Preferred
Octadecyl rhodamine B chloride is an amphiphilic fluorescent probe for staining cell membranes and investigating membrane dynamics. It associates with lipid bilayers and exhibits strong fluorescence as a monomer; in aqueous media it can form aggregates that are blue-shifted and lead to self-quenching. It is used in membrane biology, cell labeling, and fluorescence-based assays.
Catalog No. 50-002-82714