Learn More
ALADDIN SCIENTIFIC CORPORATION OctadecyltrimethoxysilaneODTMS, 25ml
Supplier:  ALADDIN SCIENTIFIC CORPORATION T10656225ML
Encompass_Preferred
Trimethoxy(octadecyl)silane (OTS) is an important alkylsilane that can form strong covalent bonds with silicon oxide and serves as a potential lubricant for micro electro mechanical systems (MEMS) OTS is largely used for surface chemical modification modifying silica resulted in an increased charge carrier mobility for a variety of vacuum deposited and solution cast organic semiconductors For preparing hydrophobic coatings and self-assembled monolayers
Catalog No. 50-000-82582