Learn More
Medchemexpress LLC Octaethylene glycol monomethyl ether | 25990-96-9 | MFCD06797152 | 99.8% | 384.46 g/mol | C17H36O9 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0426571G
Encompass_Preferred
Octaethylene glycol monomethyl ether is a methyl-terminated polyethylene glycol (PEG8) linker used in the synthesis of PROTACs and other bioconjugates. It is a colorless liquid with CAS 25990-96-9, molecular formula C17H36O9, and molecular weight 384.46 g/mol. Typical purity is 99.84% and it is commonly supplied in 1 g packaging.
  • Flexible, hydrophilic spacer for linker construction.
  • Methyl-terminated ether group for stable end-capping.
  • High purity suitable for synthetic applications.
  • Provides analytical documentation such as COA, HNMR, and MS.
  • Usable as a PEG-based building block in medicinal chemistry and bioconjugation.
Catalog No. 50-002-10234