Learn More
AdipoGen Octafluoronaphthalene
SDP
Supplier:  AdipoGen CDXO0054G005
Encompass_Preferred
Chemical. CAS 313-72-4. Formula C10F8. MW 272.09. Aryl fluorinated building block for synthesis or intermediate in chemical synthesis. Used in the synthesis of agrochemical, pharmaceutical and fluorescent dye field. Also used as a standard for evaluating the sensitivity of GC-MS devices.
Catalog No. 50-256-0316