Learn More
Cowie Technology Corporation OCTAHEDRAL STIR 38MM L 8MM D
Supplier:  Cowie Technology Corporation 001.538Y
Encompass_Preferred
This item has a minimum qty of 41 per supplier requirements.
Stirbar, octahedral, length 38 mm, diameter 8 mm
Catalog No. 50-823-153