Learn More
Medchemexpress LLC Octahydropyrrolo[2,1-b][1,3]oxazepine-7-carboxamide derivative | 1422180-49-1 | MFCD34469389; MFCD01632616 | 98.4% | 456.58 g/mol | C25H36N4O4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12537850MG
Encompass_Preferred
SBP-0636457 is a SMAC mimetic research compound used to investigate apoptosis and cancer biology in preclinical and in vitro studies. The compound is supplied as a solid powder with high reported purity, is typically dissolved in DMSO for stock solutions, and is available in multiple milligram sizes for assay flexibility. CAS number 1422180-49-1 identifies the substance unambiguously.
Catalog No. 50-002-37887