Learn More
Medchemexpress LLC Octamethylcyclotetra 50g
SDP
Supplier:  Medchemexpress LLC HYW11600750G
Encompass_Preferred
Octamethylcyclotetrasiloxane promotes the anchorage-independent growth of MCF-10A and MCF-10F cells Octamethylcyclotetrasiloxane induces DNA damage inhibits the expression of DNA-repairing protein BRCA1 under long-term and high-concentration exposure Octamethylcyclotetrasiloxane exhibits intrinsic estrogenic activity[1]
Catalog No. 50-002-06680