Learn More
Medchemexpress LLC Octan-2-yl 2-cyanoac 25mg | 133978-15-1 | 209.28 g/mol | C12H19NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11654425MG
Encompass_Preferred
Octan-2-yl 2-cyanoacrylate is a cyanoacrylate ester supplied for research and industrial use; it functions as a fast-curing adhesive and tissue adhesive, polymerizing rapidly in the presence of moisture to form strong bonds.
  • High purity (98.0%) suitable for research applications.
  • Low-viscosity, colorless to light yellow liquid for easy application.
  • Soluble in DMSO (~16.67 mg/mL) with warming or sonication recommended.
  • Store under nitrogen at 4°C; long-term storage in solvent at -80°C.
  • Used for tissue adhesion, wound closure, and industrial bonding.
Catalog No. 50-003-08269