Learn More
Medchemexpress LLC Octanedioic-2,2,3,3,4,4,5,5,6,6,7,7-d12 acid | 169397-99-3 | 98.6% | 186.27 g/mol | C8H2D12O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015300S1100MG
Encompass_Preferred
Suberic acid-d12 is the deuterium-labeled isotopologue of suberic (octanedioic) acid supplied for use as an internal standard, tracer, or reagent in metabolic, biochemical, and mass spectrometry studies of fatty acid metabolism and related enzyme deficiencies.
  • Deuterium-labeled tracer for accurate quantitation by mass spectrometry.
  • High chemical purity (98.6%).
  • Known molecular weight and formula for analytical reference: 186.27 g/mol, C8H2D12O4.
  • Multiple pack sizes available to support method development and scaling.
  • Storage recommendations provided for powder and solution to preserve stability.
Catalog No. 50-003-02477