Learn More
Medchemexpress LLC Octanoic acid, 4-ethyl- | 16493-80-4 | MFCD00506494 | 99.1% | 172.26 g/mol | C10H20O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0153075G
Encompass_Preferred
4-Ethyloctanoic acid (octanoic acid, 4-ethyl-, CAS 16493-80-4) is a naturally occurring fatty acid derivative first isolated from Saussurea lappa Clarke. It is used in food chemistry and flavor research and is supplied at high purity for analytical and preparative studies.
  • High purity suitable for analytical and research applications.
  • Natural product origin documented from Saussurea lappa.
  • Functions as a flavoring compound and food additive in studies.
  • Chemical identifiers: C10H20O2, mol wt 172.26 g/mol, CAS 16493-80-4.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-003-02412