Learn More
Medchemexpress LLC Octanoic acid, 4-ethyl- | 16493-80-4 | MFCD00506494 | 99.1% | 172.27 g/mol | C10H20O2 | 10 G

Supplier: Medchemexpress LLC HYW01530710G
4-Ethyloctanoic acid is a naturally occurring branched medium-chain carboxylic acid first isolated from Saussurea lappa Clarke. It is used as a food flavoring compound and additive and is supplied for research and analytical applications.
- High reported purity: 99.05%.
- Available in multiple sizes, including 10 g.
- Certificate of Analysis and safety data sheet available.
- Suitable as an analytical reference for flavor and formulation research.
- Molecular formula C10H20O2; molecular weight 172.27 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.